4-Amino-4'-dimethylaminoazobenzene 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: N,N-Dimethyl-4,4'-azodianiline

379298-1G
379298-1GEA 136 EUR
379298-1G
4-Amino-4'-dimethylaminoazobenzene 97%, Sigma-Aldrich®
4-Amino-4'-dimethylaminoazobenzene

N,N-Dimethyl-4,4′-azodianiline, also knows as Disperse Black 3 is a disperse azo dye. It has been used in a condensation reaction for the systhesis of azo-based phenylthiophene Schiff bases.

Formel: C₁₄H₁₆N₄
Molecular Weight: 240,31 g/mol
Schmelzpunkt: 190 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00010204
CAS-Nummer: 539-17-3
EINECS: 208-712-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=290572680ca4e9a26b889fdbbb1dd257aaad1147&vid=290572680ca4e9a26b889fdbbb1dd257aaad1147&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Orange
Appearance (Form), Powder, Crystals and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Carbon 67.5 - 72.4%
Nitrogen 22.5 - 24.1%
Purity (HPLC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR