Lithium-tert-butanolat 2.2 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

379311000 379318000
379311000.EA 95.4 EUR
379311000. 379318000.
Lithium-tert-butanolat 2.2 M in THF, AcroSeal®
Lithium-tert-butanolat
Formel: C₄H₉LiO
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00050479
CAS-Nummer: 1907-33-1
EINECS: 217-611-5
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Solution
Strength 19 to 21 %
Impurity ≤0.2 % LiOH + Li2CO3 (as LiOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR