Lithium isopropoxide 1 M in Hexanen, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

380201000
380201000.EA 83.7 EUR
380201000.
Lithium isopropoxide 1 M in Hexanen, AcroSeal®
Lithium isopropoxide
Formel: C₃H₇LiO
Molecular Weight: 66,028 g/mol
Dichte: 0,69 g/cm³
Flammpunkt: –27 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00048286
CAS-Nummer: 2388-10-5
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to orange Solution
Titration with HClO4 0.95 to 1.15 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR