di-Natriumhydrogenphosphat, anhydrous 98,0-100,5 Dried USP, multi-compendial, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: di-Natriumorthophosphat, sek. Natriumphosphat, Natriumorthophosphat dibasisch

M.O.C. Category R

3804-01EA 101.57 EUR
3804-01 3804-07 3804-65
di-Natriumhydrogenphosphat, anhydrous 98,0-100,5 Dried USP, multi-compendial, J.T.Baker®
di-Natriumhydrogenphosphat

GMP Manufactured Product Meets E.P. Chemical Specifications Meets JPE Specifications Meets U.S.P Requirements CAUTION: For Manufacturing, processing or repackaging Bulk Pharmaceutical Chemical


  • GMP manufactured product

Formel: Na₂HPO₄
Molecular Weight: 141.96 g/mol
Schmelzpunkt: 250 °C
Dichte: 2.07 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003496
CAS-Nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Bestellen

Spezifikation Testergebnisse

Meets JPE Specifications 98.0 - 100.5%
Meets E.P. Chemical Specifications Passes Test
Meets U.S.P Requirements ≤5.0%
GMP Manufactured Product ≤0.4%
CAUTION: For Manufacturing, processing or repackaging ≤0.06%
Bulk Pharmaceutical Chemical ≤0.2%
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Identification Passes Test
USP - Loss on Drying at 130°C ≤ 5.0 %
USP - Insoluble Substances ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.06 %
USP - Sulfate (SO₄) ≤ 0.2 %
USP - Arsenic (As) ≤ 16 ppm
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
JPE - Assay (dried basis) ≥ 99.0 %
JPE - Identification Passes Test
JPE - Arsenic (As) ≤ 2 ppm
JPE - Carbonate Passes Test
JPE - Chloride (Cl) ≤ 0.011 %
JPE - Clarity and Color of Solution Passes Test
JPE - Heavy Metals ≤ 10 ppm
JPE - pH (1-100) 9.0 - 9.4
JPE - Sulfate (SO₄) ≤ 0.038 %
JPE - Loss on Drying ≤ 2.0 %
EP - Assay (Na₂HPO₄) (dried basis) 98.0 - 101.0 %
EP - Appearance of Solution Passes Test
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Identification C Passes Test
EP - Identification D Passes Test
EP - Reducing Substances Passes Test
EP - Chloride (Cl) ≤ 200 ppm
EP - Iron (Fe) ≤ 20 ppm
EP - Loss on Drying ≤ 1.0 %
EP - Monosodium Phosphate ≤ 2.5 %
EP - Sulfate (SO₄) ≤ 500 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR