Lithium (trimethylsilyl)acetylide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

380651000 380658000
380651000.EA 165 EUR
380651000. 380658000.
Lithium (trimethylsilyl)acetylide 0.5 M in THF, AcroSeal®
Lithium (trimethylsilyl)acetylide
Formel: C₅H₉LiSi
Dichte: 0,88 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00075059
CAS-Nummer: 54655-07-1
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to orange Solution
Titration with HClO4 0.40 to 0.60 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR