Natriumdihydrogenphosphat Monohydrat 98,0-103,0% (durch Trockensubstanz) USP, ACS, J.T.Baker®
Lieferant: Avantor
Synonyme:
Natriumphosphat monobasisch Monohydrat
3808-65EA
0
EUR
3808-65
Natriumdihydrogenphosphat Monohydrat 98,0-103,0% (durch Trockensubstanz) USP, ACS, J.T.Baker®
Natriumdihydrogenphosphat Monohydrat
Formel:
NaH₂PO₄·H₂O Molecular Weight: 137,99 g/mol Schmelzpunkt: 100 °C Dichte: 2,04 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149208 CAS-Nummer: 10049-21-5 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 13,08734 |
Spezifikation Testergebnisse
GMP Manufactured Product | 98.0 - 103.0 % |
Meets ACS Reagent Chemical Requirements | Passes Test |
Meets U.S.P Requirements |
|
CAUTION: For Manufacturing, processing or repackaging |
|
Bulk Pharmaceutical Chemical |
|
USP - Assay (anhydrous basis) | 98.0 - 103.0 % |
USP - Aluminum, Calcium, and Related Elements | Passes Test |
USP - Arsenic (As) | ≤8 ppm |
USP - Chloride (Cl) | ≤0.014 % |
USP - Identification | Passes Test |
USP - Insoluble Substances | ≤0.2 % |
USP - pH (1 in 20) | 4.1 - 4.5 |
USP - Sulfate (SO₄) | ≤0.15 % |
USP - Water (H₂O) | 10.0 - 15.0 % |
Assay (NaH₂PO₄·H₂O) | 98.0 - 102.0 % |
pH of 5% Solution at 25°C | 4.1 - 4.5 |
Insoluble Matter | ≤0.01 % |
Chloride (Cl) | ≤5 ppm |
ACS - Sulfate (SO₄) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Heavy Metals (as Pb) | ≤0.001 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
For EP notated requirements, results are derived from testing Sodium | |
Phosphate, Monobasic Monohydrate, U.S.P. to the EP Chemical | |
specifications for Sodium Dihydrogen Phosphate Dihydrate. | |
For JPE notated requirements, results are derived from testing Sodium | |
Phosphate, Monobasic Monohydrate, U.S.P. to the JPE Chemical | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...