Sudan Black B Staining System, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
MQ500
Intended for the histochemical demonstration of neutrophil granules in blood or bone marrow films.
- Suitable for haematology
- Suitable for histology
Principle
Following fixation, blood or bone marrow films are immersed in a buffered Sudan Black B solution. After rinsing, slides are counterstained with Gill hematoxylin, and cells are examined microscopically for the presence of blue-black discrete granulation. Granulocytic precursors demonstrate groups of coarse blue-black deposits. Other hematopoietic cells are typically negative.
Kit component also available separately
Glutaraldehyde solution, for in vitro diagnostic use 75 mL (3802)
Kit component only
Sudan Black B Staining Reagent (3801) 200 mL
Hematoxylin Solution, Gill No. 3 (GHS3) 100 mL
Lagertemperatur:
Kühlschrank |
UN:
3316 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...