Magnetic beads, Sera-Mag Oligo (dT) coated magnetic particles
Lieferant: Cytiva
38152103011150 38152103010150 38152103010350
38152103011150.EA 143.88 EUR
38152103011150. 38152103010150. 38152103010350.
Magnetic beads, Sera-Mag Oligo (dT) coated magnetic particles
Kugeln Magnetkugeln
Sera-Mag Oligo(dT)-Coated Magnetic Particles enable convenient mRNA isolation and extraction. Sera-Mag Oligo(dT) Magnetic Particles bind target mRNA through pairing of the polyadenylated RNA tail found on the 3' end of mRNA to the covalently bound oligo(dT) groups on the surface of the particles. This binding is easily accomplished using standard hybridisation conditions. Versatile and efficient methods for mRNA purification are critical, given that eukaryotic mRNA makes up only 1 to 3% of total cellular RNA. Sera-Mag Oligo(dT) magnetic particles remove 90% or more of mRNA from total RNAMore Product Information
Bestellen
TECHNISCHE DATEN
Bindungskapazität 11 µg mRNA/mg beads
Environmentally Preferable
Particle size nominal 1 µm diameter