di-Natriumhydrogenphosphat Heptahydrat 98,0-100,5% (Trockenmasse), Kristalle USP Endotoxin getested, J.T.Baker®

Lieferant: Avantor
Achtung

3816-07EA 1067.35 EUR
3816-07 3816-05
di-Natriumhydrogenphosphat Heptahydrat 98,0-100,5% (Trockenmasse), Kristalle USP Endotoxin getested, J.T.Baker®
di-Natriumhydrogenphosphat Heptahydrat
Formel: HNa₂O₄P·7H₂O
Schmelzpunkt: 48 °C
Dichte: 1,7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149180
CAS-Nummer: 7782-85-6
EINECS: 231-448-7

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤16 ppm
CAUTION: For Manufacturing, processing or repackaging ≤0.06%
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 16 ppm
USP - Chloride (Cl) ≤ 0.06 %
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
USP - Identification Passes Test
USP - Insoluble Substances ≤ 0.4 %
USP - Loss on Drying 43.0 - 50.0 %
USP - Sulfate (SO₄) ≤ 0.2 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR