Natriumdihydrogenphosphat Dihydrat 98,0-102,0%, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

3819-01EA 194.77 EUR
3819-01
Natriumdihydrogenphosphat Dihydrat 98,0-102,0%, BAKER ANALYZED®, J.T.Baker®
Natriumdihydrogenphosphat Dihydrat

Lot analysis on label.

Formel: NaH₂PO₄·2H₂O
Siedepunkt: ? 650 °C (1013 hPa)
Schmelzpunkt: 60 °C
Dichte: 1,92 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149209
CAS-Nummer: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 98.0 - 102.0%
Assay (NaH₂PO₄·2H₂O) 98.0 - 102.0 %
pH of 5% Solution at 25°C 4.1 - 4.5
Chloride (Cl) ≤5 ppm
Insoluble Matter, Calcium and NH₄OH Precipitate ≤0.01 %
Nitrogen Compounds (as N) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
Arsenic (As) ≤0.5 ppm
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR