Natriumhydroxid concentrate for dilution to 1 L, 0.5 M NaOH (0.5 N) Volumetric, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Caustic soda

38217-1EA
38217-1EAEA 50.33 EUR
38217-1EA
Natriumhydroxid concentrate for dilution to 1 L, 0.5 M NaOH (0.5 N) Volumetric, Fluka™
Sodium hydroxide

Concentrate for 1 L standard solution

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 120 °C
Schmelzpunkt: 323
Dichte: 1.350 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR