di-Natriumhydrogenphosphat Dodecahydrat 98,0-102,0%, Kristalle, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

3822-07EA 0 EUR
3822-07
di-Natriumhydrogenphosphat Dodecahydrat 98,0-102,0%, Kristalle, BAKER ANALYZED®, J.T.Baker®
di-Natriumhydrogenphosphat Dodecahydrat

Crystals. Lot analysis on label.

Formel: Na₂HPO₄·12H₂O
Schmelzpunkt: 35 °C
Dichte: 1,52 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149181
CAS-Nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 98.0 - 102.0%
Assay 98.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 8.7 - 9.3
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Nitrogen Compounds (as N) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR