Natriumthiosulfat concentrate for dilution to 1L, 0.01 M Na2S2O3 (0.01N) Volumetric, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
38243-1EA
38243-6X1EA
38243-1EAEA
57.34
EUR
38243-1EA
38243-6X1EA
Natriumthiosulfat concentrate for dilution to 1L, 0.01 M Na2S2O3 (0.01N) Volumetric, Fluka™
Natriumthiosulfat
Concentrate for 1L standard solution
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 °C (1.013 hPa) Schmelzpunkt: 48 Dichte: 1.0 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 Merck Index: 12,08844 |
Spezifikation Testergebnisse
Factor | 0.998 - 1.002 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...