di-Natriumhydrogenphosphat, wasserfrei ≥98% USP, ACS Endotoxin getested, J.T.Baker®

Lieferant: Avantor


Veränderungsmanagement-Kategorie= HR

3826-07EA 952.71 EUR
3826-07
di-Natriumhydrogenphosphat, wasserfrei ≥98% USP, ACS Endotoxin getested, J.T.Baker®
di-Natriumhydrogenphosphat
Formel: Na₂HPO₄
Schmelzpunkt: 250 °C
Dichte: 1,53 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003496
CAS-Nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements 98.0 - 100.5%
GMP Manufactured Product ≤3 ppm
Meets ACS Reagent Chemical Requirements ≤0.06%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.2%
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Chloride (Cl) ≤ 0.06 %
USP - Identification Passes Test
USP - Insoluble Matter ≤ 0.2 %
USP - Loss on Drying at 130°C ≤ 5.0 %
USP - Sulfate (SO₄) ≤ 0.2 %
ACS - Assay (Na₂HPO₄) (by acidimetry) ≥ 99.0 %
ACS - Chloride (Cl) ≤ 0.002 %
ACS - Heavy Metals (as Pb) ≤ 0.001 %
ACS - Insoluble Matter ≤ 0.01 %
ACS - Iron (Fe) ≤ 0.001 %
ACS - Loss on Drying at 105°C ≤ 0.2 %
ACS - pH of 5% Solution at 25°C 8.7 - 9.3
ACS - Sulfate (SO₄) ≤ 0.005 %
Endotoxin Concentration (5 EU/g max.) Passes Test
(Suitable for Buffer Solutions) ≤5.0%
No Class 1,2,3 or other solvents are used or produced in the Passes Test
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR