di-Natriumhydrogenphosphat, wasserfrei ≥99,0% (durch Acidimetrie), BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

3828-07EA 626.48 EUR
3828-07 3828-01
di-Natriumhydrogenphosphat, wasserfrei ≥99,0% (durch Acidimetrie), BAKER ANALYZED® ACS, J.T.Baker®
di-Natriumhydrogenphosphat
Formel: Na₂HPO₄
Schmelzpunkt: 250 °C
Dichte: 2,07 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003496
CAS-Nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.01%
Assay (Na₂HPO₄) (by acidimetry) ≥99.0 %
Insoluble Matter ≤0.01 %
Loss on Drying at 105°C ≤0.2 %
pH of 5% Solution at 25°C 8.7 - 9.3
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR