di-Natriumhydrogenphosphat, anhydrous 99,0 Acidimetry, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Achtung
|
Synonyme:
di-Natriumorthophosphat, sek. Natriumphosphat, Natriumorthophosphat dibasisch
3828-07EA
626.48
EUR
3828-07
3828-01
di-Natriumhydrogenphosphat, anhydrous 99,0 Acidimetry, BAKER ANALYZED® ACS, J.T.Baker®
di-Natriumhydrogenphosphat
Formel:
Na₂HPO₄ Molecular Weight: 141.96 g/mol Schmelzpunkt: 250 Dichte: 2.07 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003496 CAS-Nummer: 7558-79-4 EINECS: 231-448-7 REACH: 01-2119489797-11 Merck Index: 12,08805 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | ≥99.0% |
For Laboratory,Research,or Manufacturing Use | ≤0.01% |
Assay (Na₂HPO₄) (by acidimetry) | ≥99.0 % |
Insoluble Matter | ≤0.01 % |
Loss on Drying at 105°C | ≤0.2 % |
pH of 5% Solution at 25°C | 8.7 - 9.3 |
Chloride (Cl) | ≤0.002 % |
Sulfate (SO₄) | ≤0.005 % |
Heavy Metals (as Pb) | ≤0.001 % |
Iron (Fe) | ≤0.002 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...