Silbernitrat concentrate for dilution to 1L, 0.5 M AgNO3 (0.5N) Volumetric, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Höllenstein
38311-1EA
38311-1EAEA
234.82
EUR
38311-1EA
Silbernitrat concentrate for dilution to 1L, 0.5 M AgNO3 (0.5N) Volumetric, Fluka™
Silbernitrat
Concentrate for 1L standard solution
Formel:
AgNO₃ Molecular Weight: 169.87 g/mol Siedepunkt: 100 °C Schmelzpunkt: 210 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003414 CAS-Nummer: 7761-88-8 EINECS: 231-853-9 UN: 3264 ADR: 8 II Merck Index: 12,08661 |
|
Spezifikation Testergebnisse
Factor | 0.998 - 1.002 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...