tri-Natriumphosphat Dodecahydrat 98,0-102,0% (durch Acidimetrie), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Natriumphosphat tribasisch Dodecahydrat

3836-07EA 0 EUR
3836-07
tri-Natriumphosphat Dodecahydrat 98,0-102,0% (durch Acidimetrie), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
tri-Natriumphosphat Dodecahydrat

Crystals. Lot analysis on label.

Formel: Na₃PO₄·12H₂O
Schmelzpunkt: ∼75 °C
Dichte: 1,62 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149198
CAS-Nummer: 10101-89-0
EINECS: 231-509-8
UN: 3262
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use ≤2.5%
Assay (Na₃PO₄·12H₂O) (by acidimetry) 98.0 - 102.0 %
Excess Alkali (as NaOH) ≤2.5 %
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR