Cerium(III) ammonium nitrate tetrahydrate, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
38367.22
38367.36
38367.22EA
65.9
EUR
38367.22
38367.36
Cerium(III) ammonium nitrate tetrahydrate, Reagenzqualität
Cerium(III) ammonium nitrate tetrahydrate
CAS No.: 13083-04-0
Molecular Formula: H8CeN7O15.4H2O
Formula Weight: 558.28 (486.22anhy)
Physical Form: Crystalline
Appearance: White
UN No.: UN1477
MDL No.: MFCD00210608
Formel:
H₈CeN₇O₁₅·4H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210608 CAS-Nummer: 13083-04-0 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...