Ethyl viologen diperchlorate, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
1,1'-Diethyl-4,4'-bipyridinium diperchlorate
384100-5G
384100-5GEA
226
EUR
384100-5G
Ethyl viologen diperchlorate, Sigma-Aldrich®
Ethyl viologen diperchlorate
Ethyl viologen diperchlorate has been used in in-situ spectroscopic studies of viologen based electrochromic device.
Formel:
C₁₄H₁₈Cl₂N₂O₈ Schmelzpunkt: 270 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134591 CAS-Nummer: 36305-51-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...