Vanadium ≥99,5% (Metall-Basis), Drahtform, Ø 2.0 mm (0.08 in)

Lieferant: Thermo Fisher Scientific

38516.G1
38516.G1EA 306 EUR
38516.G1
Vanadium ≥99,5% (Metall-Basis), Drahtform, Ø 2.0 mm (0.08 in)
Vanadium

Purity: 99.5% CAS No.: 7440-62-2 Physical Form: ≈19.2g/m Odour: Odourless MDL No.: MFCD00011453

Formel: V
Siedepunkt: 3477 °C (1013 hPa)
Schmelzpunkt: 1896 °C
Dichte: 6,015 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011453
CAS-Nummer: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR