Aluminiumtrifluormethansulfonat
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)
38568.09
38568.09EA
98.8
EUR
38568.09
Aluminiumtrifluormethansulfonat
Aluminiumtrifluormethansulfonat
CAS No.: 74974-61-1
Molecular Formula: C3AlF9O9S3
Formula Weight: 474.20
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: ca 300°
UN No.: 3261
MDL No.: MFCD00143596
Formel:
C₃AlF₉O₉S₃ Molecular Weight: 474,19 g/mol Schmelzpunkt: ∼ 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143596 CAS-Nummer: 74974-61-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...