Certetrafluorid Hydrat
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cer(IV)fluorid Hydrat
38580.09
38580.09EA
78.1
EUR
38580.09
Certetrafluorid Hydrat
Certetrafluorid Hydrat
CAS No.: 60627-09-0
Molecular Formula: CeF4.xH2O
Formula Weight: 216.11(anhy)
Physical Form: Powder
Solubility: Insoluble in water. Slowly hydrolyzed by cold water
UN No.: 3288
MDL No.: MFCD00049422
Formel:
CeF₄·H₂O Molecular Weight: 234,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049422 CAS-Nummer: 60627-09-0 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...