Magnesium(II) phthalocyanine ≥90%, tech.

Lieferant: Thermo Fisher Scientific

38601.03 38601.06
38601.03EA 55.4 EUR
38601.03 38601.06
Magnesium(II) phthalocyanine ≥90%, tech.

Purity: 90% CAS No.: 1661-03-6 Molecular Formula: C32H16MgN8 Formula Weight: 536.84 Physical Form: Powder MDL No.: MFCD00041953

Formel: C₃₂H₁₆MgN₈
Lagertemperatur: Raumtemperatur
MDL: MFCD00041953
CAS-Nummer: 1661-03-6
EINECS: 216-766-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR