Bis(pentamethylcyclopentadienyl)mangan(II) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Decamethylmanganocene, Bis(pentamethylcyclopentadienyl)manganese
38605.03
38605.03EA
110
EUR
38605.03
Bis(pentamethylcyclopentadienyl)mangan(II) ≥98%
Bis(pentamethylcyclopentadienyl)mangan(II)
Purity: 98%
CAS No.: 67506-86-9
Molecular Formula: C20H30Mn
Formula Weight: 325.38
Physical Form: Crystalline, Packaged in ampoules
Sensitivity: Air and Moisture Sensitive
MDL No.: MFCD00058844
Formel:
C₂₀H₃₀Mn Schmelzpunkt: 230 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058844 CAS-Nummer: 67506-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...