Citrate Buffer, Concentrate, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
3861-20ML
3861-20MLEA 131 EUR
3861-20ML
Citrate Buffer, Concentrate, Sigma-Aldrich®
Puffer Biologische Puffer
Citrate buffer solution, 0.38 mol/L, pH 5.4 when diluted according to procedure 386A.
  • For in vitro diagnostic use