Hydrocortison, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Cortisol, 11β,17α,21-Trihydroxy-4-pregnen-3,20-dion, 11β,17α,21-Trihydroxypregn-4-en-3,20-dion, 11β-Hydroxycortison

3867-1
3867-1EA 84.7 EUR
3867-1
Hydrocortison, Millipore®
Hydrocortison
Formel: C₂₁H₃₀O₅
Molecular Weight: 362.47 g/mol
Siedepunkt: 567
Schmelzpunkt: 202…204 °C
Dichte: 1.4 g/cm³ (20 °C)
MDL: MFCD00011654
CAS-Nummer: 50-23-7
EINECS: 200-020-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR