Methylorange 0,1% (w/v) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
Synonyme:
Goldorange, Helianthin, Natrium-4-(4-dimethylaminophenylazo)benzolsulfonat, Orange III, 4-(4-(Dimethylamino)phenylazo)benzolsulfonsäure Natriumsalz, (4-(Dimethylamino)azobenzol-4'-sulfonsäure Natriumsalz
38695.AP
38695.APEA
40.7
EUR
38695.AP
Methylorange 0,1% (w/v) in wässriger Lösung
Methylorange
CAS No.: 547-58-0
Molecular Formula: C14H14N3NaO3S
Formula Weight: 327.34
Physical Form: Liquid
Density: 0.987
Odour: Odourless
MDL No.: MFCD00007502
Formel:
C₁₄H₁₄N₃NaO₃S Dichte: 0,987 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007502 CAS-Nummer: 547-58-0 EINECS: 208-925-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...