Acid Phosphatase, Leukocyte (TRAP) Kit Select reagents packaged in gelatin capsules, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ500

386A-1KT
386A-1KTEA 374 EUR
386A-1KT
Acid Phosphatase, Leukocyte (TRAP) Kit Select reagents packaged in gelatin capsules, Sigma-Aldrich®
Acid Phosphatase, Leukocyte (TRAP) Kit

Kit is used to demonstrate acid phosphatase and tartrate resistant acid phosphatase (TRAP) in blood, bone marrow and tissue touch preparations.


  • Suitable for haematology
  • Suitable for histology


Peripheral blood or bone marrow preparations are fixed to a microscope slide. The resulting film is incubated in a solution of Naphthol AS-BI phosphoric acid and Fast Garnet GBC.


Kit component only
Acetate Solution (386-3) 50 mL
Acid Hematoxylin (285-2) 100 mL
Citrate Concentrate (386-1) 20 mL
Fast Garnet GBC Salt (kit only) 2 × 10 caps
Naphthol AS-BI Phosphoric Acid (386-4) 40 mL
Tartrate Solution (kit only) 20 mL

Lagertemperatur: Kühlschrank
UN: 3316
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR