Dithiothreitol (DTT, Cleland's Reagenz) ≥99,4%, OmniPur® Calbiochem®, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
DTT, (R*,R*)-1,4-Dimercapto-2,3-butandiol
3870-25GM
3870-25GMEA
304
EUR
3870-25GM
Dithiothreitol (DTT, Cleland's Reagenz) ≥99,4%, OmniPur® Calbiochem®, Millipore®
Dithiothreitol (DTT, Cleland's Reagenz)
Used for reduction of protein disulfide linkages and in molecular biology.
Bitte beachten Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Formel:
C₄H₁₀O₂S₂ Molecular Weight: 154.25 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00004877 CAS-Nummer: 3483-12-3 |
|
Spezifikation Testergebnisse
A (0.02 M in water) | ≤0.05 AU |
Assay (-SH content) | ≥99.4% |
DNAse | None Detected |
Infrared Spectrum | Conforms to Standard |
Loss on Drying | ≤0.5% |
Melting Point | 39 - 43 °C |
Protease | None Detected |
RNAse | None Detected |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...