Phenolphthalein ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
38705.22
38705.36
38705.22EA
37.1
EUR
38705.22
38705.36
Phenolphthalein ACS
Phenolphthalein
CAS No.: 77-09-8
Molecular Formula: C20H14O4
Formula Weight: 318.33
Physical Form: Powder
Density: 1.299
Melting Point: 258-262°
Beilstein Reference: 284423
Merck Reference: 14,7243
MDL No.: MFCD00005913
Indicator
Formel:
C₂₀H₁₄O₄ Schmelzpunkt: 258…262 °C Dichte: 1,299 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005913 CAS-Nummer: 77-09-8 EINECS: 201-004-7 REACH: 01-2119498295-24 Merck Index: 14,7243 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...