Alizaringelb R (Natriumsalz)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
38707.09
38707.14
38707.09EA
65.2
EUR
38707.09
38707.14
Alizaringelb R (Natriumsalz)
Alizaringelb R (Natriumsalz)
CAS No.: 1718-34-9
Molecular Formula: C13H8N3NaO5
Formula Weight: 309.22
Physical Form: Powder
Appearance: Dark yellow to brown
Odour: Odourless
Beilstein Reference: 962365
Merck Reference: 14,255
MDL No.: MFCD00067121
Formel:
C₁₃H₈N₃NaO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067121 CAS-Nummer: 1718-34-9 EINECS: 217-002-4 Merck Index: 14,255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...