Fast Garnet GBC Base-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
3872-10ML
3872-10MLEA
195
EUR
3872-10ML
Fast Garnet GBC Base-Lösung, Sigma-Aldrich®
Fast Garnet GBC Base
Intended for use in Sigma Procedure 387 for the demonstration of Acid Phosphatase (TRAP).
Lagertemperatur:
Kühlschrank |
MDL:
MFCD00007733 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...