Hämatoxylin-Lösung Gill II, J.T.Baker®

Lieferant: Avantor

Synonyme: Hydroxybrasilin, Naturschwarz 1

3873.2500EA 80.22 EUR
3873.2500
Hämatoxylin-Lösung Gill II, J.T.Baker®
Hämatoxylin-Färbelösung

Intended for histology and cytology staining procedures.


Hematoxyline (papanicolaou I), modified to match gill II and harris.

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
MDL: MFCD00078111
CAS-Nummer: 517-28-2
IVDLogo

Bestellen

Spezifikation Testergebnisse

Performance Test Passes Test
For Laboratory, Research or Manufacturing Use

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR