Acid Phosphatase, Leukocyte (TRAP) Kit Kit formulated with all liquid reagents, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ600
Kit is used to demonstrate acid phosphatase and tartrate resistant acid phosphatase (TRAP) in blood, bone marrow and tissue touch preparations.
- Suitable for haematology
- Suitable for histology
Demonstration of lymphocyte acid phosphatase (focal staining pattern) in cytocentrifuge or buffy coat preparations from blood, bone marrow film and tissue touch preparations typifies T-lymphocytes.
Kit component also available separately
Acetate solution, 2.5 M, pH 5.2 50 mL (3863)
Citrate Solution, pH 3.6±0.1 (25 °C), 27 mM 50 mL (915)
Fast Garnet GBC Base solution 10 mL (3872)
Naphthol AS-BI Phosphoric acid solution 10 mL (3871)
Sodium nitrite solution 10 mL (914)
Tartrate Solution 10 mL (3873)
Kit component only
Hematoxylin Solution, Gill No. 3 (kit only) 50 mL
Lagertemperatur:
Kühlschrank |
UN:
3316 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...