N,N-Dimethylacetamid 99,5 GC_ASSAY_METHOD, Puriss. p.a., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Essigsäuredimethylamid

38840-2.5L 38840-1L 38840-50L
38840-2.5LEA 182.63 EUR
38840-2.5L 38840-1L HONC38840-50L
N,N-Dimethylacetamid 99,5 GC_ASSAY_METHOD, Puriss. p.a., Riedel-de Haën™
N,N-Dimethylacetamid

Beilstein Number: 1737614

Formel: CH₃CON(CH₃)₂
Molecular Weight: 87.12 g/mol
Siedepunkt: 164…166 °C (1.013 hPa)
Schmelzpunkt: –20 °C
Dichte: 0.94 g/cm³ (20 °C)
Flammpunkt: 11.5 %(V) (99 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008686
CAS-Nummer: 127-19-5
EINECS: 204-826-4
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay (GC) Min. 99.5 %
Refractive index (n 20/D) 1.436 - 1.439
Colour of the solution complying
Turbidity of solution complying
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.008 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR