Ammoniak 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

388441000
388441000.EA 109 EUR
388441000.
Ammoniak 0.5 M in THF, AcroSeal®
Ammonia
Formel: NH₃
Dichte: 0,85 g/cm³
Flammpunkt: –36 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011418
CAS-Nummer: 7664-41-7
UN: 1993
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Solution
Water ≤2 % (K.F.)
Strength 0.40 to 0.60 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR