Malachitgrün-Oxalat-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

38978-100ML-F
38978-100ML-FEA 39.6 EUR
38978-100ML-F
Malachitgrün-Oxalat-Lösung, Sigma-Aldrich®
Malachitgrün-Oxalat

Staining of bacterial spores according to Schaeffer and Fulton; Pappenheim stain when combined with Gram′s stain

Formel: C₅₂H₅₄N₄O₁₂
MDL: MFCD00011766
CAS-Nummer: 2437-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7723ac34e7273345d2b069c0836679df388eaaf7&vid=ec3b640e950ddc753e47d460407fdd3277490d06&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d712338fae1d16ee9b5bb9fef6ee138153de6b&vid=b1bcce866dc7e726ed2770ec2ea1047580498ba4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR