N-Methyl-2-pyrrolidon (NMP) ≥99,5% für die HPLC
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
38986.K2
38986.K7
38986.M1
38986.K2EA
132
EUR
38986.K2
38986.K7
38986.M1
N-Methyl-2-pyrrolidon (NMP) ≥99,5% für die HPLC
N-Methyl-2-pyrrolidon (NMP)
Purity: 99.5%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formel:
C₅H₉NO Molecular Weight: 99.13 g/mol Siedepunkt: 204 °C Schmelzpunkt: –24 °C Dichte: 1.033 Flammpunkt: 86 °C (187 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003193 CAS-Nummer: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...