2-(Ethylthio)benzothiazole ≥97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

389900-5G
389900-5GEA 131 EUR
389900-5G
2-(Ethylthio)benzothiazole ≥97%, Sigma-Aldrich®
2-(Ethylthio)benzothiazole

2-(Ethylthio)benzothiazole,97%

Formel: C₉H₉NS₂
Molecular Weight: 195,3 g/mol
Dichte: 1,226 g/mL (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00192166
CAS-Nummer: 2757-92-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3553d9d597af452edcf651a373520f47745a2633&vid=91c258abab7fce2980b4e4245f76b27be5272c35&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR