Cooled Incubators, Series 4
Lieferant: LMS
1200 600A 1200A 600NP 1200NP
390-0083EA 13670 EUR
390-0083 390-0168 390-0210 390-0100 390-0084
Cooled Incubators, Series 4
Brutschränke Kühlbrutschränke
LMS Series 4 cooled incubators are temperature controlled cabinets with fan assisted air circulation via a pre-mixing chamber. The units have a stainless steel exterior, stainless steel lined interior with polyurethane foam insulation, and feature a door lock and magnetic door gasket. Corrosion resistant inner chamber is easy to clean, disinfection procedures are detailed in the user manual.
  • Units feature automatic defrost
  • Easy to use controls with digital display of actual temperature (and set point for programmable models)
  • Hermetically sealed
More Product Information