Heracell Vios CR CO₂ incubators, CTS series, Thermo Scientific™
Lieferant: Thermo Scientific
51033772 51033775 51033784 51033787
390-1576EA 20200 EUR
390-1576 390-1577 390-1578 390-1579
Heracell Vios CR CO₂ incubators, CTS series, Thermo Scientific™
Brutschränke CO₂-Brutschränke
Heracell Vios CR cleanroom compatible CO₂ incubators are third party-certified, suitable for use in ISO Class 5 and GMP Grade A/B environments.
  • Next level particulate and contamination control
  • Active particle control system captures particulates to help assure continued cleanroom compatibility
  • Sealed exterior casing with stainless steel interior and exterior
  • IP 54 rated electronics, protected from dust and spray in any direction
  • Thermo Scientific™ THRIVE™ active airflow technology delivers homogeneous growth conditions quickly
  • IR sensor and proven 180 °C high heat sterilisation
  • Documentation to support the validation process
  • 4 to 20 mA data output included for external data collection
  • Choice of two sizes - 165 and 255 L
Vios CR third party certified, cleanroom-compatible CO₂ incubators have been designed to actively control particle emissions to meet ISO Class 5 and GMP Grade A/B conditions. These incubators have been enhanced to improve compatibility with common cleanroom cleaning procedures and include all the necessary documentation to support cell therapy production in a cleanroom setting. As expected of the Thermo Scientific™ Heracell™ Vios™ CO₂ incubator family, this incubator continues to offer a superior incubation environment.
Standards: Third party validated by TUV SUD as appropriate for use in ISO Class 5 and GMP grade A/B environments, in accordance with ISO 14644-1. Safety cabinets by TÜV Nord.
Bestellinformation: Compatible accessories are available and must be ordered separately: Lower profile roller base, floor stand and stacking adapter plate – these are all TUV SUD approved and available in both sizes.