3,3′-Diethylthiacyanine iodide ~97% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

390410-1G
390410-1GEA 285 EUR
390410-1G
3,3′-Diethylthiacyanine iodide ~97% (Farbstoffgehalt), Sigma-Aldrich®
3,3′-Diethylthiacyanine iodide

3,3′-Diethylthiacyanine iodide (DTTHCI) is a fluorophore. 3,3′-Diethylthiacyanine iodide has been used to investigate the kinetics of fluorescence staining in bacteria (two Gram-negative and two Gram-positive species).

Formel: C₁₉H₁₉IN₂S₂
Molecular Weight: 466,40 g/mol
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00050309
CAS-Nummer: 2197-01-5
EINECS: 218-593-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2540ce2ca4c5d320892c08d9153671e94aace87&vid=f2540ce2ca4c5d320892c08d9153671e94aace87&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour), Yellow to Dark Green Conforms to Requirements
Appearance (Form), Powder or Crystals Conforms to Requirement
Infrared spectrum Conforms to Structure
Wavelength, 0.004 g/L, Ethanol 420 - 428 nm
Extinction Coefficient ≥65000
Carbon 47.2 - 50.6%
Nitrogen 5.7 - 6.2%
Purity (HPLC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR