UltraClear® Histology, J.T.Baker®

Lieferant: Avantor
Gefahr

3905.9010PEEA 65.8 EUR
3905.9010PE 3905.5000PE 3905.2500PE
UltraClear® Histology, J.T.Baker®
UltraClear®

Clearing agent for use in Histology and Cytology.

Flammpunkt: 5 °C
Lagertemperatur: Raumtemperatur
EINECS: 292-459-0
UN: 3295
ADR: 3 III
IVDLogo

Bestellen

Spezifikation Testergebnisse

Appearance passes test
Aromates (as Xylene) max. 60 ppm
Solubility in Ethanol passes test
Solubility in Xylene passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR