Hexan (Isomerengemisch) zur Analyse
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
390740010
390740025
390740100
390740010.EA
32.1
EUR
390740010.
390740025.
390740100.
Hexan (Isomerengemisch) zur Analyse
Hexan (Isomerengemisch)
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 69 °C (760 mmHg) Schmelzpunkt: –95 °C Dichte: 0,659 g/cm³ Flammpunkt: –22 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 92112-69-1 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Spezifikation Testergebnisse
Appearance | Clear liquid |
Water | ≤0.01 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Acidity | ≤0.0002 meq/g |
Substance darkened by H2SO4 | Passes test |
Aluminium (Al) | ≤0.5 ppm |
Barium (Ba) | ≤0.1 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Sulfur compounds (as S) | ≤0.005 % |
Aromatics | ≤0.01 % Benzene |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...