5-(4-(Dimethylamino)benzyliden)rhodanin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

39090-10G
39090-10GEA 91.6 EUR
39090-10G
5-(4-(Dimethylamino)benzyliden)rhodanin, Supelco®
5-(4-(Dimethylamino)benzyliden)rhodanin
Formel: C₁₂H₁₂N₂OS₂
MDL: MFCD00064857
CAS-Nummer: 536-17-4
EINECS: 208-625-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cece0e9e2759437a207b7b3b6c8986ca4f747b1a&vid=7efcfa96dfcb3c9386db662d5784876a7148afa2&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR