Peroxidase (Myeloperoxidase) Leukocyte Kit Hanker-Yates substrate kit, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ500
390A-1KT
390A-1KTEA
350
EUR
390A-1KT
Peroxidase (Myeloperoxidase) Leukocyte Kit Hanker-Yates substrate kit, Sigma-Aldrich®
Peroxidase (Myeloperoxidase) Leukocyte Kit
Staining system for polymorphonucleocytes in blood or bone marrow films.
- Suitable for haematology
- Suitable for histology
- Sufficient for 10-120 manual assays
The staining characteristics of polymorphonucleocytes are used to distinguish acute myelocytic leukemia from other types of leukemia.
Kit component also available separately
Trizmal™ buffer, pH 6.3(concentrate), 0.2 M 50 mL (903C)
Kit component only
Acid Hematoxylin Solution (285-2) 100 mL
Peroxidase Indicator Reagent (390-1) 10 vial(s)
Lagertemperatur:
Kühlschrank |
MDL:
MFCD00131641 UN: 3316 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...