Petri dish holder
Lieferant: SciLabware
SWP062
391-1150EA 0 EUR
391-1150
Petri dish holder
Gestelle Spender für Petrischalen
Polyamide epoxy coated steel. Holds up to six 90 mm petri dishes.
  • Better heat and chemical resistance than other coatings
  • Ideal for small batch incubation