Stopfen, mit umstülpbarem Rand, Versilic®
Lieferant: Saint Gobain Life Sciences
467007-100 467010-50 467015-20 467020-20 467025-10 467030-05 467018-20 467013-50
391-2260EA 110 EUR
391-2260 217-0187 217-0189 217-0191 391-2261 391-2262 217-0190 217-0188
Stopfen, mit umstülpbarem Rand, Versilic®
Stopfen Silikonstopfen
These stoppers are made of translucent silicone. The stoppers guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers are imperfect.
  • Autoclavable
  • Outstanding sealing and protection from the outside
  • Biologically neutral, good chemical resistance
  • Resistant to ageing, ozone and repeated sterilisation
  • Temperature resistant from –60 to +220 °C (up to +250 °C for short periods)
The stoppers can be punctured and resealed after sampling, even after several uses.
Resistant to temperatures from -60 to +220 °C (up to +250 °C for short periods)