Racks for centrifuge tubes, PP
Lieferant: GLW
G23 G26 G18
391-8848EA 22.5 EUR
391-8848 391-8849 GLWAG18
Racks for centrifuge tubes, PP
Gestelle Reagenzglasgestelle
  • Unique screw assembly provides superior durability and assured stability
  • Space efficient designed that is for use in the safety cabinet
  • Use in freezer down to −90 °C, autoclavable up to +121 °C